Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_012390.3 | NP_036522.3 | 3 | 348 | |
XM_017008021.1 | XP_016863510.1 | 2 | 319 | ||
GenBank mRNA | AK312075.1 | - | 3 | 363 | |
BC140927.1 | - | 2 | 294 | 9056739 | |
CD722957.1 | - | 3 | 348 |