Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_022582.2 | NP_072104.2 | 3 | 400 | |
XM_006228668.2 | XP_006228730.1 | 6 | 658 | ||
XM_006228670.2 | XP_006228732.1 | 5 | 830 | ||
GenBank mRNA | AF036941.1 | - | 3 | 409 | |
U67883.1 | - | 4 | 409 |