Gene Transcripts

Gene Symbol
Gpc1
Entrez Gene ID
58920
Gene Name
glypican 1
Gene Aliases
HSPG M12, HSPGM12
Chromosome Location
Chr. 9: 99998275 - 100026818 on Build Rnor_6.0
UniGene ID
Rn.7044
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030828.1 NP_110455.1 3 625
GenBank mRNA BC061572.1 - 3 623
L02896.1 - 3 625
L34067.1 - 3 458

View Full Product Details