Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_138508.4 | NP_612517.2 | 4, 5 | 357 | |
GenBank mRNA | BC081713.1 | - | 4, 5 | 370 | |
M57453.1 | - | 4, 5 | 357 | ||
M58287.1 | - | 3, 4 | 243 | ||
M62763.1 | - | 4, 5 | 384 | ||
X60654.1 | - | 4, 5 | 337 |