Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_012633.2 | NP_036765.2 | 1 | 328 | |
XM_006257321.2 | XP_006257383.1 | 1 | 1111 | ||
XM_006257322.2 | XP_006257384.1 | 1 | 1109 | ||
XM_008765669.1 | XP_008763891.1 | 1 | 1113 | ||
GenBank mRNA | AF031878.1 | - | 1 | 321 | |
BC100656.1 | - | 1 | 328 | 7936222 |