Gene Transcripts

Gene Symbol
Serpina3c
Entrez Gene ID
24794
Gene Name
serine (or cysteine) proteinase inhibitor, clade A, member 3C
Gene Aliases
CPi-21, GHR-P63, Kbp, Klkbp, SPI-2, SPI-2.3, SPI2, Serpina3k, Spin2, Spin2b
Chromosome Location
Chr. 6: 127996162 - 128003435 on Build Rnor_6.0
UniGene ID
Rn.91257
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012657.2 NP_036789.2 5 1228
XM_006240458.2 XP_006240520.1 5 1271
GenBank mRNA BC062236.1 - 5 1232
D00751.1 - 5 1181
FQ209380.1 - 5 1231
FQ209577.1 - 5 1230
FQ209663.1 - 4 956
FQ210286.1 - 5 1234
FQ218085.1 - 5 1233
FQ218647.1 - 4 905
FQ218802.1 - 5 1230
FQ219296.1 - 5 1234
FQ219312.1 - 5 1232
FQ219600.1 - 5 1228
M14320.1 - 4 726
M15916.1 - 5 1193
X05348.1 - 5 1215
X16358.1 - 5 1229

View Full Product Details