Gene Transcripts

Gene Symbol
ICMT
Entrez Gene ID
23463
Gene Name
isoprenylcysteine carboxyl methyltransferase
Gene Aliases
HSTE14, MST098, MSTP098, PCCMT, PCMT, PPMT
Chromosome Location
Chr. 1: 6221193 - 6235984 on Build GRCh38
UniGene ID
Hs.515688
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012405.3 NP_036537.1 5 864
XM_011541140.2 XP_011539442.1 4 564
GenBank mRNA AF064084.1 - 5 834
AK092890.1 - 6 805
BC017037.1 - 6 959
BC028168.1 - 5 820 5245125
CR541671.1 - 5 791
CR541711.1 - 5 791

View Full Product Details