Gene Transcripts

Gene Symbol
Cckar
Entrez Gene ID
24889
Gene Name
cholecystokinin A receptor
Gene Aliases
Cck-ar
Chromosome Location
Chr. 14: 59610951 - 59621856 on Build Rnor_6.0
UniGene ID
Rn.10184
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012688.3 NP_036820.1 2 393
XM_008770188.1 XP_008768410.1 3 577
GenBank mRNA M88096.1 - 2 397

View Full Product Details