Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001284413.1 | NP_001271342.1 | 6 | 850 | |
NM_001284415.1 | NP_001271344.1 | 6 | 853 | ||
NM_012260.3 | NP_036392.2 | 7 | 931 | ||
NR_104315.1 | 6 | 858 | |||
GenBank mRNA | AF161397.1 | - | 7 | 551 | |
AJ131753.1 | - | 7 | 641 | ||
AK301546.1 | - | 6 | 567 | ||
BC001627.1 | - | 7 | 649 | 3538159 | |
BC007440.2 | - | 6 | 563 |