Gene Transcripts
- Gene Symbol
- Gata1
- Entrez Gene ID
- 25172
- Gene Name
- GATA binding protein 1
- Gene Aliases
- Eryf1
- Chromosome Location
- Chr. X: 15374750 - 15382572 on Build Rnor_6.0
- UniGene ID
- Rn.10024
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_012764.1 |
NP_036896.1
|
|
1368 |
|
GenBank mRNA |
D13518.1 |
- |
|
1368 |
|
View Full Product Details