Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001142418.1 | NP_001135890.1 | 5 | 1881 | |
NM_001142419.1 | NP_001135891.1 | 4 | 1814 | ||
NM_001142420.1 | NP_001135892.1 | 3 | 1661 | ||
NM_001142421.1 | NP_001135893.1 | 4 | 1763 | ||
NM_001142422.1 | NP_001135894.1 | 5 | 1863 | ||
NM_001142423.1 | NP_001135895.1 | 4 | 1781 | ||
NM_001142424.1 | NP_001135896.1 | 5 | 1830 | ||
NM_001142425.1 | NP_001135897.1 | 4 | 1801 | ||
NM_001142426.1 | NP_001135898.1 | 3 | 1679 | ||
NM_001142427.1 | NP_001135899.1 | 4 | 1783 | ||
NM_001142428.1 | NP_001135900.1 | 5 | 1812 | ||
NM_001142429.1 | NP_001135901.1 | 5 | 1850 | ||
NM_001142430.1 | NP_001135902.1 | 4 | 1728 | ||
NM_001142431.1 | NP_001135903.1 | 4 | 1710 | ||
NM_001142432.1 | NP_001135904.1 | 5 | 1832 | ||
NM_012286.2 | NP_036418.1 | 4 | 1832 | ||
GenBank mRNA | AB050779.1 | - | 1 | 260 | |
AF100620.1 | - | 4 | 1680 | ||
AF167174.1 | - | 4 | 1675 | ||
AK056012.1 | - | 4 | 1670 | ||
AK095298.1 | - | 2 | 2592 | ||
AK126115.1 | - | 6 | 1659 | ||
AK126427.1 | - | 4 | 2608 | ||
BC056899.1 | - | 5 | 1666 | 3905425 | |
BC093013.1 | - | 5 | 1711 | 30528536 | |
D14812.1 | - | 4 | 1680 |