Gene Transcripts

Gene Symbol
Gusb
Entrez Gene ID
24434
Gene Name
glucuronidase, beta
Gene Aliases
Ac2-223, Gus-s
Chromosome Location
Chr. 12: 30202066 - 30215583 on Build Rnor_6.0
UniGene ID
Rn.3692
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017015.2 NP_058711.2 11 1761
GenBank mRNA AY321342.1 - 1894
M13962.1 - 11 1731
Y00717.1 - 11 1687

View Full Product Details