Gene Transcripts

Gene Symbol
Pgm1
Entrez Gene ID
24645
Gene Name
phosphoglucomutase 1
Gene Aliases
Chromosome Location
Chr. 5: 118743632 - 118803055 on Build Rnor_6.0
UniGene ID
Rn.9970
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017033.2 NP_058729.2 6 1024
XM_006238442.2 XP_006238504.1 7 1215
XM_008763853.1 XP_008762075.1 6 1567
GenBank mRNA BC099807.1 - 6 1024
BC128703.1 - 6 1029
L11694.1 - 6 1007

View Full Product Details