Gene Transcripts
- Gene Symbol
- Slc4a3
- Entrez Gene ID
- 24781
- Gene Name
- solute carrier family 4 (anion exchanger), member 3
- Gene Aliases
- Ae3, Aep3
- Chromosome Location
- Chr. 9: 82743028 - 82758213 on Build Rnor_6.0
- UniGene ID
- Rn.87739
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_017049.1 |
NP_058745.1
|
3, 4 |
532 |
|
GenBank mRNA |
J05167.1 |
- |
3, 4 |
532 |
|
View Full Product Details