Gene Transcripts
- Gene Symbol
- Tacr3
- Entrez Gene ID
- 24808
- Gene Name
- tachykinin receptor 3
- Gene Aliases
- Nkr, Nmkr, Tac3r
- Chromosome Location
- Chr. 2: 240021152 - 240118971 on Build Rnor_6.0
- UniGene ID
- Rn.9702
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_017053.1 |
NP_058749.1
|
1 |
346 |
|
GenBank mRNA |
J05189.1 |
- |
1 |
346 |
|
View Full Product Details