Gene Transcripts

Gene Symbol
Myo5b
Entrez Gene ID
25132
Gene Name
myosin Vb
Gene Aliases
Myr6
Chromosome Location
Chr. 18: 70427103 - 70729985 on Build Rnor_6.0
UniGene ID
Rn.10640
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017083.1 NP_058779.1 2 282
XM_008772085.1 XP_008770307.1 2 245
XM_008772086.1 XP_008770308.1 2 245
GenBank mRNA U60416.1 - 2 282

View Full Product Details