Gene Transcripts
- Gene Symbol
- Pgam2
- Entrez Gene ID
- 24959
- Gene Name
- phosphoglycerate mutase 2 (muscle)
- Gene Aliases
- D14Mgh1, PGAM-M, Pgmut
- Chromosome Location
- Chr. 14: 86045005 - 86047116 on Build Rnor_6.0
- UniGene ID
- Rn.9738
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_017328.2 |
NP_059024.1
|
2 |
590 |
|
GenBank mRNA |
M31835.1 |
- |
2 |
591 |
|
View Full Product Details