Gene Transcripts

Gene Symbol
Psg19
Entrez Gene ID
24256
Gene Name
pregnancy specific glycoprotein 19
Gene Aliases
CEAC, Cea3, Cgm3, RATCEAC
Chromosome Location
Chr. 1: 79680194 - 79690460 on Build Rnor_6.0
UniGene ID
Rn.100787
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001270654.1 NP_001257583.1 3 792
NM_019126.2 NP_061999.2 3 792
GenBank mRNA BC097328.1 - 3 766
U09815.1 - 792

View Full Product Details