Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_012392.3 | NP_036524.1 | 5 | 1458 | |
NR_033686.1 | 4 | 1157 | |||
NR_033688.1 | 5 | 1299 | |||
XM_011541745.1 | XP_011540047.1 | 5 | 1085 | ||
XM_011541746.2 | XP_011540048.1 | 5 | 1552 | ||
XM_011541747.1 | XP_011540049.1 | 5 | 1427 | ||
XM_017001680.1 | XP_016857169.1 | 5 | 1416 | ||
XM_017001681.1 | XP_016857170.1 | 5 | 2723 | ||
GenBank mRNA | AB018357.1 | - | 5 | 1097 | |
AB026628.1 | - | 5 | 1097 | ||
AK001420.1 | - | 5 | 1110 | ||
AK092483.1 | - | 1 | 2452 | ||
AY359011.1 | - | 5 | 1101 | ||
BC002773.2 | - | 5 | 1087 | 3615612 | |
BC012561.1 | - | 5 | 1103 | 4273213 |