Gene Transcripts

Gene Symbol
Wdr7
Entrez Gene ID
66031
Gene Name
WD repeat domain 7
Gene Aliases
Trag, Wd7
Chromosome Location
Chr. 18: 59096640 - 59396312 on Build Rnor_6.0
UniGene ID
Rn.211985
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023975.1 NP_076465.1 2 180
XM_008772171.1 XP_008770393.1 3 387
GenBank mRNA AF192379.1 - 2 180
AF305813.1 - 2 180

View Full Product Details