Gene Transcripts

Gene Symbol
PLXNB2
Entrez Gene ID
23654
Gene Name
plexin B2
Gene Aliases
MM1, Nbla00445, PLEXB2, dJ402G11.3
Chromosome Location
Chr. 22: 50274979 - 50307637 on Build GRCh38
UniGene ID
Hs.3989
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012401.3 NP_036533.2 27 4568
XM_005261909.1 XP_005261966.1 28 4616
XM_005261910.2 XP_005261967.1 28 4627
XM_005261911.4 XP_005261968.1 27 4566
XM_006724413.2 XP_006724476.1 29 4686
XM_011530682.2 XP_011528984.1 28 4767
XM_017028703.1 XP_016884192.1 29 4667
XM_017028704.1 XP_016884193.1 28 4620
GenBank mRNA AB002313.1 - 26 4485
AB075511.1 - 12 1678
AK025701.1 - 14 1936
AK056543.1 - 10 1737
AK074932.1 - 9 1204
AK126394.1 - 8 1277
BC004542.2 - 4 387
BC148225.1 - 5 747
BC148280.1 - 5 747
BT006887.1 - 4 371
S76730.1 - 5 720

View Full Product Details