Gene Transcripts

Gene Symbol
Aard
Entrez Gene ID
246323
Gene Name
alanine and arginine rich domain containing protein
Gene Aliases
A5D3
Chromosome Location
Chr. 7: 91588458 - 91593299 on Build Rnor_6.0
UniGene ID
Rn.38451
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145093.1 NP_659561.1 2 513
GenBank mRNA AY007690.1 - 2 513

View Full Product Details