Gene Transcripts

Gene Symbol
Sh2d2a
Entrez Gene ID
310688
Gene Name
SH2 domain containing 2A
Gene Aliases
Chromosome Location
Chr. 2: 187218851 - 187225000 on Build Rnor_6.0
UniGene ID
Rn.9034
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_207605.3 NP_997488.2 7 1204
GenBank mRNA AF468787.2 - 7 1196
BC088087.1 - 7 1204 7379832

View Full Product Details