Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001282186.1 | NP_001269115.1 | 2 | 1218 | |
NM_001282187.1 | NP_001269116.1 | 4 | 1388 | ||
NM_001282188.1 | NP_001269117.1 | 4 | 1365 | ||
NM_006639.3 | NP_006630.1 | 3 | 1305 | ||
GenBank mRNA | AF119711.1 | - | 1 | 804 | |
AF133266.1 | - | 1 | 804 | ||
AK313643.1 | - | 3 | 1229 | ||
BC035750.1 | - | 3 | 1097 | 5750221 |