Gene Transcripts
- Gene Symbol
- RXFP1
- Entrez Gene ID
- 59350
- Gene Name
- relaxin/insulin like family peptide receptor 1
- Gene Aliases
- LGR7, RXFPR1
- Chromosome Location
- Chr. 4: 158521711 - 158653372 on Build GRCh38
- UniGene ID
- Hs.591686
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001253727.1 |
NP_001240656.1
|
13, 14 |
1398 |
|
NM_001253728.1 |
NP_001240657.1
|
12, 13 |
1218 |
|
NM_001253729.1 |
NP_001240658.1
|
11, 12 |
1173 |
|
NM_001253732.1 |
NP_001240661.1
|
14, 15 |
1401 |
|
NM_001253733.1 |
NP_001240662.1
|
13, 14 |
1329 |
|
NM_021634.3 |
NP_067647.2
|
13, 14 |
1317 |
|
NR_045579.1 |
|
13, 14 |
1934 |
|
NR_045580.1 |
|
13, 14 |
1370 |
|
NR_045581.1 |
|
13, 14 |
1341 |
|
NR_045582.1 |
|
11, 12 |
1278 |
|
NR_045584.1 |
|
14, 15 |
1370 |
|
XM_011532175.1 |
XP_011530477.1
|
12, 13 |
1295 |
|
XM_011532176.1 |
XP_011530478.1
|
12, 13 |
1217 |
|
XM_011532179.2 |
XP_011530481.1
|
13, 14 |
1367 |
|
XM_017008519.1 |
XP_016864008.1
|
14, 15 |
1331 |
|
XM_017008520.1 |
XP_016864009.1
|
13, 14 |
1420 |
|
XM_017008521.1 |
XP_016864010.1
|
14, 15 |
1250 |
|
XM_017008523.1 |
XP_016864012.1
|
13, 14 |
1289 |
|
XM_017008524.1 |
XP_016864013.1
|
12, 13 |
1217 |
|
XM_017008525.1 |
XP_016864014.1
|
12, 13 |
1221 |
|
XM_017008526.1 |
XP_016864015.1
|
8, 9 |
788 |
|
GenBank mRNA |
AB208943.1 |
- |
14, 15 |
1139 |
|
AF190500.1 |
- |
13, 14 |
1054 |
|
AK122647.1 |
- |
15, 16 |
1358 |
|
AK294694.1 |
- |
13, 14 |
1162 |
|
AK295040.1 |
- |
13, 14 |
1757 |
|
AK300379.1 |
- |
12, 13 |
1113 |
|
AK312470.1 |
- |
13, 14 |
1161 |
|
AY899848.1 |
- |
14, 15 |
1107 |
|
AY899849.1 |
- |
11, 12 |
910 |
|
AY899850.1 |
- |
12, 13 |
955 |
|
BC112142.1 |
- |
13, 14 |
1136 |
8327610 |
BC113617.1 |
- |
13, 14 |
1139 |
8322669 |
BC143556.1 |
- |
13, 14 |
1159 |
|
BC143558.1 |
- |
13, 14 |
1192 |
|
BX647975.1 |
- |
13, 14 |
1255 |
|
BX647980.1 |
- |
13, 14 |
1255 |
|
BX647985.1 |
- |
13, 14 |
1284 |
|
View Full Product Details