Gene Transcripts

Gene Symbol
TAS2R43
Entrez Gene ID
259289
Gene Name
taste 2 receptor member 43
Gene Aliases
T2R43, T2R52
Chromosome Location
Chr. 12: 11091287 - 11092313 on Build GRCh38
UniGene ID
Hs.688195
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_176884.2 NP_795365.2 1 664
GenBank mRNA BC117423.1 - 1 663

View Full Product Details