Gene Transcripts

Gene Symbol
POLL
Entrez Gene ID
27343
Gene Name
polymerase (DNA) lambda
Gene Aliases
BETAN, POLKAPPA
Chromosome Location
Chr. 10: 101578882 - 101588270 on Build GRCh38
UniGene ID
Hs.523230
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001174084.1 NP_001167555.1 9 2174
NM_001174085.1 NP_001167556.1 9 1835
NM_001308382.1 NP_001295311.1 6 1016
NM_013274.3 NP_037406.1 9 2111
NR_033406.1 9 1710
XM_006717775.1 XP_006717838.1 6 1779
XM_006717776.1 XP_006717839.1 7 1007
XM_006717777.1 XP_006717840.1 5 1737
XM_011539650.1 XP_011537952.1 9 1800
XM_011539651.1 XP_011537953.1 9 1779
XM_011539652.1 XP_011537954.1 9 2088
XM_011539653.1 XP_011537955.1 9 2081
XM_011539654.1 XP_011537956.1 9 1524
XM_011539655.1 XP_011537957.1 9 1503
XM_011539656.1 XP_011537958.1 8 1482
XM_011539657.1 XP_011537959.1 9 2027
XM_011539659.1 XP_011537961.1 6 2076
XM_011539660.1 XP_011537962.1 7 1332
XM_011539662.1 XP_011537964.1 6 1800
XM_011539663.1 XP_011537965.1 7 1028
XM_011539664.1 XP_011537966.1 5 1758
XM_011539665.2 XP_011537967.1 6 1346
XM_011539666.1 XP_011537968.1 6 1024
XM_011539667.1 XP_011537969.1 8 1302
XM_017016084.1 XP_016871573.1 7 1497
XM_017016085.1 XP_016871574.1 7 1476
XM_017016086.1 XP_016871575.1 7 2169
XM_017016087.1 XP_016871576.1 7 1221
XM_017016089.1 XP_016871578.1 7 1605
XM_017016090.1 XP_016871579.1 7 1472
XM_017016091.1 XP_016871580.1 6 2055
XM_017016093.1 XP_016871582.1 6 1283
GenBank mRNA AF161019.1 - 8 1740
AF218027.1 - 3 916
AF283478.1 - 8 1679
AJ131890.1 - 9 2050
AJ844641.1 - 3 617
AJ844642.1 - 2 485
AK021600.1 - 10 1663
AK022476.1 - 9 1688
AK091106.1 - 2 2294
AK094956.1 - 2 1253
AK127896.1 - 4 1446
AK128521.1 - 2 958
AK292225.1 - 9 1757
AK293421.1 - 8 1332
AK293602.1 - 7 1007
AK300862.1 - 6 1138
AY302442.1 - 8 1403
BC003548.1 - 5 1669 3536366
BC068529.1 - 9 2063 5267859

View Full Product Details