Gene Transcripts

Gene Symbol
MKRN1
Entrez Gene ID
23608
Gene Name
makorin ring finger protein 1
Gene Aliases
RNF61
Chromosome Location
Chr. 7: 140453040 - 140479569 on Build GRCh38
UniGene ID
Hs.744883
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145125.1 NP_001138597.1 5 1028
NM_001291663.1 NP_001278592.1 7 1190
NM_013446.3 NP_038474.2 5 1028
NR_117084.1 3 571
XM_011515996.1 XP_011514298.1 4 817
XM_011515997.2 XP_011514299.1 5 1085
XM_011515998.1 XP_011514300.1 5 770
GenBank mRNA AB209298.1 - 5 795
AF117233.1 - 5 958
AF192784.1 - 5 923
AK027137.1 - 4 540
AK127030.1 - 6 1645
AK297361.1 - 3 370
AK315552.1 - 5 827
AL136812.1 - 5 926
AM236048.1 - 5 809
BC025955.1 - 5 961 4869484
BC037400.1 - 5 813 5176176
BC064838.1 - 5 806 6173715

View Full Product Details