Gene Transcripts
- Gene Symbol
- CYTH4
- Entrez Gene ID
- 27128
- Gene Name
- cytohesin 4
- Gene Aliases
- CYT4, DJ63G5.1, PSCD4
- Chromosome Location
- Chr. 22: 37282383 - 37315348 on Build GRCh38
- UniGene ID
- Hs.170944
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001318024.1 |
NP_001304953.1
|
4, 5 |
448 |
|
NM_013385.4 |
NP_037517.1
|
4, 5 |
429 |
|
XM_011530147.2 |
XP_011528449.1
|
4, 5 |
316 |
|
XM_011530148.1 |
XP_011528450.1
|
4, 5 |
278 |
|
GenBank mRNA |
AF075458.1 |
- |
4, 5 |
358 |
|
AF125349.1 |
- |
4, 5 |
429 |
|
AK024428.1 |
- |
4, 5 |
283 |
|
AK315052.1 |
- |
4, 5 |
265 |
|
BC017780.1 |
- |
4, 5 |
297 |
|
BC041161.1 |
- |
4, 5 |
287 |
5745783 |
CR456551.1 |
- |
4, 5 |
278 |
|
View Full Product Details