Gene Transcripts
- Gene Symbol
- ASF1A
- Entrez Gene ID
- 25842
- Gene Name
- anti-silencing function 1A histone chaperone
- Gene Aliases
- CGI-98, CIA, HSPC146
- Chromosome Location
- Chr. 6: 118894077 - 118909171 on Build GRCh38
- UniGene ID
- Hs.292316
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_014034.2 |
NP_054753.1
|
1, 2 |
441 |
|
GenBank mRNA |
AB028628.1 |
- |
1, 2 |
267 |
|
AF151856.1 |
- |
3, 4 |
718 |
|
AF161495.1 |
- |
3, 4 |
718 |
|
AF279306.1 |
- |
1, 2 |
253 |
|
AK025738.1 |
- |
1, 2 |
298 |
|
AL050261.1 |
- |
1, 2 |
296 |
|
BC010878.1 |
- |
1, 2 |
280 |
3546858 |
CR457347.1 |
- |
1, 2 |
104 |
|
DA902777.1 |
- |
1, 2 |
441 |
|
View Full Product Details