Gene Transcripts

Gene Symbol
CHMP4A
Entrez Gene ID
29082
Gene Name
charged multivesicular body protein 4A
Gene Aliases
C14orf123, CHMP4, CHMP4B, HSPC134, SHAX2, SNF7, SNF7-1, VPS32-1, VPS32A
Chromosome Location
Chr. 14: 24209581 - 24213830 on Build GRCh38
UniGene ID
Hs.279761
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014169.3 NP_054888.2 2 537
GenBank mRNA AB100262.2 - 2 172
AF161483.1 - 2 191
AF212243.1 - 2 167
AY329084.1 - 2 146
BC010893.1 - 2 174
BC107699.1 - 2 199
BC113533.1 - 2 537 8322585
BC113535.1 - 2 537 8322587

View Full Product Details