Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_014315.2 | NP_055130.1 | 3 | 661 | |
XM_006720094.3 | XP_006720157.1 | 3 | 1141 | ||
XM_011536610.1 | XP_011534912.1 | 3 | 394 | ||
GenBank mRNA | AF113131.1 | - | 3 | 519 | |
AF244137.1 | - | 3 | 596 | ||
AK001771.1 | - | 3 | 577 | ||
AK056298.1 | - | 3 | 1122 | ||
BC002335.2 | - | 3 | 588 | 3506591 | |
BC024192.1 | - | 3 | 589 | 4819442 | |
BX161429.1 | - | 3 | 556 | ||
CF994676.1 | - | 3 | 604 | ||
CR457205.1 | - | 3 | 261 |