Gene Transcripts
- Gene Symbol
- EXOSC2
- Entrez Gene ID
- 23404
- Gene Name
- exosome component 2
- Gene Aliases
- RRP4, Rrp4p, hRrp4p, p7
- Chromosome Location
- Chr. 9: 130693760 - 130704894 on Build GRCh38
- UniGene ID
- Hs.654643
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001282708.1 |
NP_001269637.1
|
4, 5 |
392 |
|
NM_014285.6 |
NP_055100.2
|
4, 5 |
392 |
|
NR_104230.1 |
|
4, 5 |
392 |
|
XM_005272176.2 |
XP_005272233.1
|
4, 5 |
390 |
|
XM_017014558.1 |
XP_016870047.1
|
3, 4 |
924 |
|
GenBank mRNA |
AK001916.1 |
- |
4, 5 |
381 |
|
AK022460.1 |
- |
4, 5 |
378 |
|
AK090512.1 |
- |
3, 4 |
924 |
|
AK296605.1 |
- |
4, 5 |
381 |
|
AK303163.1 |
- |
4, 5 |
372 |
|
BC000747.2 |
- |
4, 5 |
362 |
2821702 |
BU934039.1 |
- |
|
395 |
|
View Full Product Details