Gene Transcripts

Gene Symbol
RTCB
Entrez Gene ID
51493
Gene Name
RNA 2',3'-cyclic phosphate and 5'-OH ligase
Gene Aliases
C22orf28, DJ149A16.6, FAAP, HSPC117
Chromosome Location
Chr. 22: 32387575 - 32412287 on Build GRCh38
UniGene ID
Hs.474643
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014306.4 NP_055121.1 2 239
GenBank mRNA AF155658.1 - 2 187
AF161466.1 - 2 198
AK297826.1 - 2 197
AK297908.1 - 199
AK301945.1 - 2 239
AK312503.1 - 2 199
AL050255.1 - 2 183
BC000151.1 - 2 177 2984824
BC002970.1 - 2 177 3533883
BC010308.1 - 2 189 3451576
BC016707.1 - 2 204 4102148
CR456450.1 - 2 154
CR457175.1 - 2 108
DC396330.1 - 2 241

View Full Product Details