Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_014315.2 | NP_055130.1 | 13 | 1630 | |
XM_011536610.1 | XP_011534912.1 | 13 | 1363 | ||
GenBank mRNA | AF113131.1 | - | 13 | 1488 | |
AF244137.1 | - | 13 | 1565 | ||
AK001771.1 | - | 13 | 1546 | ||
AK056298.1 | - | 13 | 2091 | ||
BC002335.2 | - | 13 | 1557 | 3506591 | |
BC024192.1 | - | 13 | 1558 | 4819442 | |
BX161429.1 | - | 12 | 1597 | ||
CD356831.1 | - | 3 | 291 |