Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_014358.3 | NP_055173.1 | 3, 4 | 393 | |
XM_011520614.2 | XP_011518916.1 | 2, 3 | 666 | ||
GenBank mRNA | AB024718.1 | - | 3, 4 | 379 | |
AK312671.1 | - | 3, 4 | 394 | ||
AY358499.1 | - | 3, 4 | 333 | ||
BC000715.2 | - | 3, 4 | 311 | 3507103 | |
DA678013.1 | - | 3, 4 | 394 |