Gene Transcripts

Gene Symbol
PGLYRP3
Entrez Gene ID
114771
Gene Name
peptidoglycan recognition protein 3
Gene Aliases
PGLYRPIalpha, PGRP-Ialpha, PGRPIA
Chromosome Location
Chr. 1: 153297589 - 153312975 on Build GRCh38
UniGene ID
Hs.348266
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_052891.2 NP_443123.1 4 529
XM_011509118.1 XP_011507420.1 6 958
XM_011509119.1 XP_011507421.1 5 850
XM_011509120.2 XP_011507422.1 6 736
GenBank mRNA AY035376.1 - 4 529
BC069741.1 - 4 515 7262478
BC069798.1 - 4 516 7262490
BC069801.1 - 4 516 7262502
BC128114.1 - 4 502 40112227
BC128115.1 - 4 502 40112231

View Full Product Details