Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_021161.4 | NP_066984.1 | 4 | 1088 | |
NM_138317.2 | NP_612190.1 | 4 | 1097 | ||
NM_138318.2 | NP_612191.1 | 4 | 768 | ||
XM_011536840.2 | XP_011535142.1 | 3 | 473 | ||
GenBank mRNA | AF279890.1 | - | 4 | 1103 | |
AF385399.1 | - | 4 | 769 | ||
AF385400.1 | - | 4 | 851 | ||
AK290518.1 | - | 4 | 1036 | ||
AK313499.1 | - | 4 | 1097 | ||
AK315263.1 | - | 4 | 756 | ||
BC075021.2 | - | 4 | 687 | 30915621 | |
BC075022.2 | - | 4 | 684 | 30915410 | |
EU978938.1 | - | 4 | 917 | ||
EU978939.1 | - | 4 | 770 | ||
EU978940.1 | - | 4 | 674 | ||
EU978941.1 | - | 4 | 674 |