Gene Transcripts

Gene Symbol
OR6B1
Entrez Gene ID
135946
Gene Name
olfactory receptor family 6 subfamily B member 1
Gene Aliases
OR7-3, OR7-9
Chromosome Location
Chr. 7: 144003997 - 144004932 on Build GRCh38
UniGene ID
Hs.632031
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005281.1 NP_001005281.1 1 786
GenBank mRNA BC137042.1 - 1 854
BC137047.1 - 1 854 9021044

View Full Product Details