Gene Transcripts

Gene Symbol
UBE2R2
Entrez Gene ID
54926
Gene Name
ubiquitin conjugating enzyme E2 R2
Gene Aliases
CDC34B, E2-CDC34B, UBC3B
Chromosome Location
Chr. 9: 33817164 - 33920403 on Build GRCh38
UniGene ID
Hs.643648
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017811.3 NP_060281.2 5 1792
XM_005251496.2 XP_005251553.1 4 1725
XM_011517949.1 XP_011516251.1 4 1677
XM_017014860.1 XP_016870349.1 3 1590
GenBank mRNA AK000426.1 - 5 1409
BC047584.1 - 5 1647 4827644

View Full Product Details