Gene Transcripts

Gene Symbol
CSDC2
Entrez Gene ID
27254
Gene Name
cold shock domain containing C2
Gene Aliases
PIPPIN, dJ347H13.2
Chromosome Location
Chr. 22: 41561010 - 41576666 on Build GRCh38
UniGene ID
Hs.310893
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014460.3 NP_055275.1 4 641
GenBank mRNA AB027011.1 - 3 364
AK313407.1 - 4 598
AL834417.1 - 4 526
BC067113.1 - 4 587 5759182
CR456457.1 - 3 376

View Full Product Details