Gene Transcripts

Gene Symbol
ESR2
Entrez Gene ID
2100
Gene Name
estrogen receptor 2
Gene Aliases
ER-BETA, ESR-BETA, ESRB, ESTRB, Erb, NR3A2
Chromosome Location
Chr. 14: 64226712 - 64338631 on Build GRCh38
UniGene ID
Hs.660607
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001040275.1 NP_001035365.1 2 554
NM_001214902.1 NP_001201831.1 1 176
NM_001271876.1 NP_001258805.1 1 176
NM_001271877.1 NP_001258806.1 1 176
NM_001291712.1 NP_001278641.1 7 1814
NM_001291723.1 NP_001278652.1 2 829
NM_001437.2 NP_001428.1 2 554
NR_073496.1 2 829
NR_073497.1 1 176
XM_017021079.1 XP_016876568.1 3 1218
XM_017021080.1 XP_016876569.1 3 2027
XM_017021081.1 XP_016876570.1 3 1123
XM_017021082.1 XP_016876571.1 3 975
XM_017021083.1 XP_016876572.1 2 392
XM_017021084.1 XP_016876573.1 3 1218
GenBank mRNA AB006589.1 - 7 1361
AB006590.1 - 1 184
AB209620.1 - 1 182
AF051427.1 - 2 504
AF051428.1 - 2 504
AF060555.1 - 2 556
AF124790.1 - 1 137
AK292370.1 - 2 391
BC024181.2 - 2 371 4825937
DQ777076.1 - 1 86
DQ777077.1 - 1 86
DQ838582.1 - 1 86
DQ838583.1 - 1 86
HQ692820.1 - 1 86
HQ692821.1 - 1 86
KJ534833.1 - 1 86

View Full Product Details