Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_006221.3 | NP_006212.1 | 4 | 845 | |
NR_038422.2 | 5 | 898 | |||
XM_011528068.2 | XP_011526370.1 | 6 | 836 | ||
GenBank mRNA | AK092970.1 | - | 3 | 2857 | |
AK097558.1 | - | 4 | 1381 | ||
AK291074.1 | - | 4 | 729 | ||
BC002899.1 | - | 4 | 734 | 3941595 | |
U49070.1 | - | 4 | 731 |