Gene Transcripts
- Gene Symbol
- TUBGCP4
- Entrez Gene ID
- 27229
- Gene Name
- tubulin gamma complex associated protein 4
- Gene Aliases
- 76P, GCP-4, GCP4, Grip76, MCCRP3
- Chromosome Location
- Chr. 15: 43371059 - 43409771 on Build GRCh38
- UniGene ID
- Hs.584887
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001286414.2 |
NP_001273343.1
|
8, 9 |
1193 |
|
NM_014444.4 |
NP_055259.2
|
8, 9 |
1193 |
|
XM_011521454.2 |
XP_011519756.1
|
8, 9 |
1193 |
|
XM_011521455.2 |
XP_011519757.1
|
8, 9 |
1091 |
|
XM_017022078.1 |
XP_016877567.1
|
8, 9 |
1193 |
|
GenBank mRNA |
AJ249677.1 |
- |
8, 9 |
899 |
|
AK001639.1 |
- |
4, 5 |
504 |
|
AK026551.1 |
- |
8, 9 |
1180 |
|
AK027703.1 |
- |
8, 9 |
1151 |
|
BC000966.2 |
- |
6, 7 |
653 |
3446803 |
BC009870.2 |
- |
8, 9 |
1118 |
3940649 |
BC012801.2 |
- |
8, 9 |
1137 |
2821891 |
View Full Product Details