Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001079862.2 | NP_001073331.1 | 4 | 560 | |
NM_001079863.1 | NP_001073332.1 | 4 | 680 | ||
NM_001178017.1 | NP_001171488.1 | 4 | 699 | ||
NM_001178041.2 | NP_001171512.1 | 5 | 686 | ||
NM_001178042.2 | NP_001171513.1 | 5 | 641 | ||
NM_001178043.2 | NP_001171514.1 | 4 | 590 | ||
NM_001282633.1 | NP_001269562.1 | 5 | 676 | ||
NM_001282634.1 | NP_001269563.1 | 5 | 646 | ||
NM_001282635.1 | NP_001269564.1 | 5 | 676 | ||
NM_001282636.1 | NP_001269565.1 | 4 | 590 | ||
NM_020548.7 | NP_065438.1 | 5 | 671 | ||
NR_104221.1 | 4 | 839 | |||
GenBank mRNA | BC006466.1 | - | 4 | 475 | |
BC062996.1 | - | 5 | 640 | 5168729 | |
BM768076.1 | - | 4 | 507 | ||
BP294452.1 | - | 4 | 502 | ||
BP318721.1 | - | 3 | 364 | ||
M14200.1 | - | 4 | 502 |