Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001118890.1 | NP_001112362.1 | 3 | 803 | |
NM_001243658.1 | NP_001230587.1 | 3 | 1221 | ||
NM_001243659.1 | NP_001230588.1 | 3 | 814 | ||
NM_002064.2 | NP_002055.1 | 3 | 1371 | ||
GenBank mRNA | AF069668.1 | - | 3 | 1174 | |
AK125636.1 | - | 2 | 3261 | ||
AW792936.1 | - | 1 | 25 | ||
BC005304.1 | - | 3 | 656 | 3682161 | |
BC010965.1 | - | 3 | 660 | 4333310 | |
BC106075.1 | - | 3 | 659 | 4444172 | |
D21238.1 | - | 3 | 595 | ||
X76648.1 | - | 3 | 656 |