Gene Transcripts

Gene Symbol
OR5M9
Entrez Gene ID
390162
Gene Name
olfactory receptor family 5 subfamily M member 9
Gene Aliases
OR11-190
Chromosome Location
Chr. 11: 56462469 - 56463401 on Build GRCh38
UniGene ID
Hs.553749
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004743.1 NP_001004743.1 1 74

View Full Product Details