Gene Transcripts

Gene Symbol
MT1X
Entrez Gene ID
4501
Gene Name
metallothionein 1X
Gene Aliases
MT-1l, MT1
Chromosome Location
Chr. 16: 56682470 - 56684196 on Build GRCh38
UniGene ID
Hs.374950
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005952.3 NP_005943.1 3 300
GenBank mRNA BC018190.1 - 3 285
BC032131.2 - 3 445 4053295
BC032338.1 - 3 287 5190391
CD101947.1 - 3 304
X76717.1 - 3 281

View Full Product Details