Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001136037.2 | NP_001129509.2 | 9 | 1022 | |
NM_001161403.1 | NP_001154875.1 | 8 | 956 | ||
NM_001161404.1 | NP_001154876.1 | 8 | 1246 | ||
NM_001256542.1 | NP_001243471.1 | 5 | 648 | ||
NM_017980.4 | NP_060450.2 | 8 | 1067 | ||
XM_005263710.2 | XP_005263767.1 | 7 | 739 | ||
XM_006712627.3 | XP_006712690.1 | 5 | 548 | ||
XM_011511453.1 | XP_011509755.1 | 7 | 803 | ||
XM_017004469.1 | XP_016859958.1 | 3 | 513 | ||
GenBank mRNA | AF484961.1 | - | 7 | 790 | |
AF520987.1 | - | 8 | 1411 | ||
AF527764.1 | - | 8 | 1021 | ||
AF527765.1 | - | 8 | 883 | ||
AF527766.1 | - | 8 | 923 | ||
AF527767.1 | - | 7 | 1179 | ||
AF527768.1 | - | 6 | 538 | ||
AF527769.1 | - | 4 | 612 | ||
AF527770.1 | - | 5 | 575 | ||
AK000906.1 | - | 6 | 438 | ||
AK022470.1 | - | 8 | 835 | ||
AK091652.1 | - | 6 | 1317 | ||
AK093692.1 | - | 2 | 1010 | ||
AK094954.1 | - | 7 | 1503 | ||
AK095790.1 | - | 11 | 1471 | ||
AK123014.1 | - | 4 | 3591 | ||
AK297645.1 | - | 10 | 1214 | ||
BC001370.2 | - | 1 | 122 | ||
BC065816.1 | - | 8 | 874 | 6181259 | |
BX458594.2 | - | 5 | 604 |