Gene Transcripts

Gene Symbol
TMUB1
Entrez Gene ID
83590
Gene Name
transmembrane and ubiquitin like domain containing 1
Gene Aliases
C7orf21, DULP, SB144
Chromosome Location
Chr. 7: 151081085 - 151083533 on Build GRCh38
UniGene ID
Hs.726215
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001136044.1 NP_001129516.1 3 1311
NM_031434.3 NP_113622.1 3 1774
GenBank mRNA AY037155.1 - 3 1292
AY358481.1 - 3 1270
AY603380.1 - 2 1296
BC000936.1 - 3 1311 3449979
BC033182.1 - 3 1449 4586932

View Full Product Details